Learn More
ENG Scientific Lendrum's Stain Kit
Description
- Lendrum's Stain Kit
- Made from certified dyes (when applicable)
- For inclusion bodies in sections
- Kit includes:
- Solution I - Mayer's Acid Alum Hematoxylin
- Solution II - Phloxine Solution (0.5%)
- Solution III - Tartrazine Solution (2.5%)
Specifications
Specifications
| Content And Storage | Hematoxylin, Potassium Alum, Sodium Iodate, Citric Acid, Phloxine, Ethanol, Calcium Chloride, Tartrazine |
| Packaging Type | Bottles |
| For Use With (Equipment) | Staining Equipment |
| Product Type | Staining Reagent |
| Quantity | 3 x 250 mL |
| Solution Type | Stain Kit |
| For Use With (Application) | Demonstration of intranuclear inclusion bodies in sections |
| Test Time | 5 to 10 min. |
| Form | Liquid |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.