Learn More
ENG Scientific Lugol's Iodine Solution Kit
Description
- Used primarily for the observation of protozoan cysts in wet mount preparations of fecal specimens.
- Results: Iodine enhances the nuclear structure and stains glycogen masses dark brown.
- Lugol's Iodine Solution Kit
- Includes Iodine, potassium iodide
Specifications
Specifications
| Quantity | 2 x 100 mL |
| Includes | Iodine, Potassium Iodide |
| For Use With (Application) | Observation of protozoan cyst in wet mount preparations of fecal specimen |
| Volume (Metric) | 100 mL |
| Product Type | Lugol's Iodine |
| Packaging | Bottles |
For in vitro diagnostic use only.