Learn More
ENG Scientific Lugol's Iodine Solution
Description
- Used primarily for the observation of protozoan cysts in wet mount preparations of fecal specimens.
- Results: Iodine enhances the nuclear structure and stains glycogen masses dark brown.
- Lugol's Iodine Solution Kit
- Includes Iodine, potassium iodide
- Bottles
Specifications
Specifications
| Type | Lugol's Iodine |
| Quantity | 2 x 250 mL |
| Includes | Iodine, potassium iodide |
| Format | Bottles |
| For Use With (Application) | Observation of protozoan cyst in wet mount preparations of fecal specimen |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.