Learn More
ENG Scientific Malarial Quick Stain Kit
Catalog No. ES3290
Change view
Click to view available options
Quantity:
2 x 250 mL
2 x 950 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES3290 2 x 250 mL
ES3300 2 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. ES3290 Supplier ENG Scientific Supplier No. 3290
Add to Cart
Add to Cart
This item is not returnable. View return policy
A combination of water-based buffered stains (pH range = 6.8 to 7.2) whereby the RBC's are hemolyzed allowing the malarial parasites to be stained.
  • Thick films provide a greater volume of blood and therefore, larger numbers of organisms for examination
  • Characteristic definitive morphologic criteria may be more difficult to see
  • The cytoplasm of white blood cells will stain with eosinophils predominant
  • For in-vitro diagnostic use only
Specifications
Quantity 2 x 250 mL
For Use With (Application) For in-vitro diagnostic use only
Product Type Stain
For in vitro diagnostic use only.