Learn More
ENG Scientific Malarial Wright Giemsa Stain Kit
Catalog No. ES3240
Change view
Click to view available options
Quantity:
3 x 250 mL
3 x 950 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES3240 3 x 250 mL
ES3250 3 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. ES3240 Supplier ENG Scientific Supplier No. 3240
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Easily visualize inclusions, extracellular forms and the size of red cells with this stain kit. For in vitro use only.
  • All cellular elements are stained
  • The use of a thin smear allows for easy visualization of inclusions, extracellular forms and the size of red cells
  • Their characteristic morphologies are used for differentiation
Specifications
Quantity 3 x 250 mL
For Use With (Application) For easy visualization of inclusions, extracellular forms and size of RBCs, in-vitro use only
Includes Solution I Fixative, Solution II Malarial Wright Giemsa Stain, Solution III Buffer
Packaging Bottles
Product Type Stain Kit
For in vitro diagnostic use only.