Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Malarial Wright Giemsa Stain Kit

Catalog No. p-3274906
Click to view available options
Quantity:
3 x 250 mL
3 x 950 mL
This item is not returnable. View return policy

Catalog No. ES3250

Supplier: ENG Scientific 3250

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Easily visualize inclusions, extracellular forms and the size of red cells with this stain kit. For in vitro use only.

  • All cellular elements are stained
  • The use of a thin smear allows for easy visualization of inclusions, extracellular forms and the size of red cells
  • Their characteristic morphologies are used for differentiation

Specifications

Quantity 3 x 950 mL
For Use With (Application) For easy visualization of inclusions, extracellular forms and size of RBCs, in-vitro use only
Includes Solution I Fixative, Solution II Malarial Wright Giemsa Stain, Solution III Buffer
Packaging Bottles

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.