Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Mallory's Acid Hematoxylin Stain (Phosphotungstic Acid)

Catalog No. ES8653
Change view
Click to view available options
Quantity:
1 x 250 mL
1 x 3800 mL (1 gal.)
1 x 950 mL
2 x 950 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES8653 1 x 3800 mL (1 gal.)
ES8650 1 x 250 mL
ES8652 2 x 950 mL
ES8651 1 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ES8653 Supplier ENG Scientific Supplier No. 8653
Add to Cart
Add to Cart

Routine stain method for the nervous system.

  • Demonstrate abnormal or diseased astrocytes
  • Useful in showing mitochondria, striations of cardiac and voluntary muscle, and as a counterstain for the Periodic Acid-Schiff (PAS) technique
  • Recommended staining time is 12 to 24 hours
  • Mallory's Acid Hematoxylin Stain
  • Made from certified dyes
  • Includes Hematoxylin, Phosphotungstic Acid, Potassium Permanganate

Specifications

Content And Storage Hematoxylin, Phosphotungstic Acid, Potassium Permanganate
Packaging Type Bottle
For Use With (Equipment) Staining Equipment
Product Type Hematoxylin
Quantity 1 x 3800 mL (1 gal.)
Solution Type Stain
For Use With (Application) Useful in showing mitochondria, striations of cardiac and voluntary muscle, and as a counterstain for the Periodic Acid-Schiff (PAS) technique
Test Time 12 to 24 hr.
Form Liquid

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.