Learn More
ENG Scientific Mallory's Acid Hematoxylin Stain (Phosphotungstic Acid)
Description
- Demonstrate abnormal or diseased astrocytes
- Useful in showing mitochondria, striations of cardiac and voluntary muscle, and as a counterstain for the Periodic Acid-Schiff (PAS) technique
- Recommended staining time is 12 to 24 hours
- Mallory's Acid Hematoxylin Stain
- Made from certified dyes
- Includes Hematoxylin, Phosphotungstic Acid, Potassium Permanganate
Specifications
Specifications
| Content And Storage | Hematoxylin, Phosphotungstic Acid, Potassium Permanganate |
| Packaging Type | Bottle |
| Product Type | Hematoxylin |
| Quantity | 1 x 250 mL |
| For Use With (Application) | Useful in showing mitochondria, striations of cardiac and voluntary muscle, and as a counterstain for the Periodic Acid-Schiff (PAS) technique |
| Test Time | 12 to 24 hr. |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.