Learn More
ENG Scientific Mallory's Aniline Blue Collagen Stain Kit
Description
- Mallory's Aniline Blue Collagen Stain Kit
- Demonstrates collagenous fibrils, cartilage, bone, amyloid, and nuclei
- Includes Aniline Blue, Orange G, Phosphotungstic Acid, Acid Fuchsin
- Kit includes Solution I - Aniline Blue and Solution II - 0.5% Acid Fuchsin
- Made from certified dyesIn vitro diagnostic use only
Specifications
Specifications
| Content And Storage | Aniline Blue, Orange G, Phosphotungstic Acid, Acid Fuchsin |
| For Use With (Equipment) | Staining Equipment |
| Product Type | Staining Reagent |
| Quantity | 2 x 950 mL |
| Solution Type | Stain |
| For Use With (Application) | In vitro diagnostic use only |
| Reagent Type | Acid Fuchsin Solution II (0.5%) |
| Form | Solution |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.