Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Manual Method Stain, Wright Stain

Catalog No. ES2030
Change view
Click to view available options
Quantity:
1 x 100 mL
1 x 3800 mL (1 gal.)
1 x 950 mL
2 x 250 mL
2 x 950 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES2030 1 x 3800 mL (1 gal.)
ES2012 1 x 100 mL
ES2011 2 x 250 mL
ES2013 1 x 950 mL
ES2010 2 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ES2030 Supplier ENG Scientific Supplier No. 2030
Add to Cart
Add to Cart

Using flooding technique, this solution will stain all cellular elements without precipitate or artifactural changes. Manual Method Stain (Wright or Wright Giemsa). Available with and without matching buffer solution.

  • Staining characteristics are similar to H-PACK.
  • Buffer is applied immediately after stain, reducing overall staining time.
  • Convenient flip-top cap for 250 mL and 950 mL sizes.
  • IN THE EVENT OF COLD WEATHER, allow solution to return to room temperature and SHAKE WELL BEFORE USE.
  • Contents: Wright Stain, Giemsa Stain, Methanol, Buffers, Stabilizers
  • These solutions are made from certified dyes (when applicable).

Specifications

Quantity 1 x 3800 mL (1 gal.)
Storage Requirements Room Temperature
For Use With (Application) For staining of cellular elements without precipitate or artifactural changes, in-vitro diagnostic use
Staining Time 5 to 7 min.
Packaging Bottle
Product Type Stain

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.