Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific May-Grunwald Stain Kit May-Grunwald Stain Solution I

Catalog No. ES3410
Change view
Click to view available options
Quantity:
2 x 100 mL
2 x 250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES3410 2 x 250 mL
ES3411 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES3410 Supplier ENG Scientific Supplier No. 3410
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The May-Grunwald Stain Kit May-Grunwald Stain Solution I is used for staining bone marrow cells.

  • Solution I May Grunwald Stain
  • Solution II Stock Giemsa
  • Contents: May-Grunwald Stain, Giemsa Stain, Methanol
  • Solutions made from certified dyes (when applicable)

Specifications

Quantity 2 x 250 mL
For Use With (Application) For use in May-Grunwald staining for bone marrow cells
Staining Time 3 min.
Packaging Bottles
Product Type Stain

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.