Learn More
ENG Scientific Mayer's Mucicarmine Stain
Description
- Includes Carmine, Aluminum Chloride
- Solution made from certified dyes
Specifications
Specifications
| Content And Storage | Carmine, Aluminum Chloride |
| Packaging Type | Bottle |
| For Use With (Equipment) | Staining Equipment |
| Product Type | Staining Reagent |
| Quantity | 250 mL |
| Solution Type | Stain |
| For Use With (Application) | In vitro diagnostic use only |
| Test Time | 15 min. |
| Form | Solution |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.