Learn More
ENG Scientific Mayer's Mucicarmine Stain Kit
Description
- Mayer's Mucicarmine Stain Kit
- Solution I - (Solution A - Weigert’s Hematoxylin, Solution B - Weigert’s Iron Chloride)
- Solution II - Mayer’s Mucicarmine Stain
- Solution III - Metanil Yellow Stain
- Contains: Ethanol, Hematoxylin, Ferric Chloride, HCl, Carmine, Aluminum Chloride, Metanil Yellow, Acetic Acid
- For in vitro diagnostic use only
Specifications
Specifications
| Content And Storage | Ethanol, Hematoxylin, Ferric Chloride, HCl, Carmine, Aluminum Chloride, Metanil yellow, Acetic Acid |
| Packaging Type | Bottles |
| For Use With (Equipment) | Staining Equipment |
| Product Type | Staining Reagent |
| Quantity | 4 x 250 mL |
| Solution Type | Stain Kit |
| For Use With (Application) | In vitro diagnostic use only |
| Form | Liquid |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.