Learn More
ENG Scientific Methyl Green with Pyronin Stain
Description
- Smears are brought to distilled water through alcohols.
Specifications
Specifications
| Content And Storage | Methyl Green, Pyronin, Ethanol, Glycerol, Phenol |
| Packaging Type | Pack (Bottles) |
| For Use With (Equipment) | Staining Equipment |
| Product Type | Staining Reagent |
| Quantity | 2 x 950 mL |
| Solution Type | Stain |
| Form | Solution |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.