Learn More
ENG Scientific Hydrochloric Acid Solution (2%)
Description
- Miscellaneous Solutions; Hydrochloric Acid (2%)
- Hydrochloric Acid
Specifications
Specifications
| Type | Hydrochloric Acid |
| Packaging Type | Bottle |
| Quantity | 1 x 475 mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.