Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Sodium Hydroxide Solution (6%)

Catalog No. ES9765
Change view
Click to view available options
Quantity:
1 x 19 L (5 gal.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES9765 1 x 19 L (5 gal.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES9765 Supplier ENG Scientific Supplier No. 9765
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Hydroxide Solution (6%)

  • Miscellaneous Solutions; Sodium Hydroxide (6%)

Specifications

Type Sodium Hydroxide
Packaging Type Cubetainer
Quantity 1 x 19 L (5 gal.)
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.