Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Modified Trichrome Stain Kit

Catalog No. ES6658
Change view
Click to view available options
Quantity:
2 x 250 mL
7 x 100 mL
Product Type:
Modified Trichome Stain Kit
Modified Trichrome Solution I
Modified Trichrome Solution II
Modified Trichrome Solution V
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
ES6658 2 x 250 mL Modified Trichrome Solution V
ES6644 2 x 250 mL Modified Trichrome Solution I
ES6647 2 x 250 mL Modified Trichrome Solution II
ES6640 7 x 100 mL Modified Trichome Stain Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. ES6658 Supplier ENG Scientific Supplier No. 6658
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For quick detection of Microsporidia Spores in Stool and Duodenal Aspirates. Non-evasive method for Modified Trichrome Stain Kit.

  • This stain method was designed as a simple, non-invasive technique for the rapid detection of microsporidia spores in fecal material by light microscopy.
  • The chromotrope based stain is a modification of the trichrome stain for intestinal protozoa.
  • Recommended Procedure:
    • Specimens for light microscopial examination should be prepared from a suspension of unconcentrated liquid stool (10 μL) in Sol. I (1:3 ratio).
    • Smears are prepared directly and thinly spread on a glass slide covering a 45 x 25 mm area.
    • Fix smears in Sol. II for 5 minutes.
    • Stain smears with Sol. III for 90 minutes.
    • Rinse slides in Sol. IV for 10 seconds.
    • Rinse briefly in Sol. V to remove acid.
    • Dehydrate in Sol. V for 5 minutes, Sol. VI for 10 minutes and Sol. VII for 10 minutes.
    • Read microscopically under oil immersion lens (magnification x 1000).
  • Results:
    • Enterocytozoon bienuesi spores appear ovoid and refractile at approximately 1.5 by 0.90 μm.
    • The spore wall of properly stained smears should appear a bright pinkish red.
    • Most bacteria and background debris stain a faint green.

Specifications

Quantity 2 x 250 mL
For Use With (Application) For the rapid detection of microsporidia spores in fecal material and duodenal aspirates by light microscopy
Product Type Modified Trichrome Solution V

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.