Learn More
ENG Scientific Nuclear Fast Red Stain (Kemechtrot)
Description
- May be used as a general counterstain in the following procedures:
- Gridley's reticulum stain
- Snook's reticulum stain
- Lepehne's method for hemoglobin
- Ralph's method for hemoglobin
- Kossa's method for calcium
- Pizzolato's method
- Puchtler-Sweat method for basement membranes
- Osmium tetroxide method for fat in frozen sections
- Gomori's method for iron
- Mallory's method for iron
- Perl's method for iron
- Tumbull's Blue method for hemosiderin
- Sudan Black B stain for fat
- Alcian Blue (short method) with and without hyaluronidase
- Mallory's reaction for iron
- Stain's bile pigment stain
- Nuclear Fast Red Stain (Kernechtrot)
- For in vitro diagnostic use only
Specifications
Specifications
| Content And Storage | Nuclear Fast Red, Aluminum Sulfate |
| Packaging Type | Bottle |
| For Use With (Equipment) | Staining Equipment |
| Product Type | Staining Reagent |
| Quantity | 1 x 250 mL |
| Solution Type | Stain |
| For Use With (Application) | Procedures like gridley reticulum stain, snook reticulum stain, lepehne method for Hemoglobin |
| Form | Liquid |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.