Learn More
ENG Scientific Page-Green Method Stain Kit
Description
- Page-Green Method Stain Kit
- For staining of distemper inclusion bodies
- Kit includes: Solution I - Harris' Alum Hematoxylin, Solution II - Acid Alcohol (1%), and Solution III - Shorr-Single Differential Stain
- Made from certified dyes (when applicable)
- For in vitro diagnostic use only
Specifications
Specifications
| Content And Storage | Hematoxylin, Potassium Alum, Ethanol, HCl, Biebrich Scarlet, Orange G, Fast Green FCF, Phosphotungstic Acid, Phosphomolybdic Acid, Glacial Acetic Acid |
| For Use With (Equipment) | Staining Equipment |
| Product Type | Staining Reagent |
| Quantity | 3 x 250 mL |
| Solution Type | Stain Kit |
| For Use With (Application) | Staining of distemper inclusion bodies |
| Form | Liquid |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.