Learn More
ENG Scientific Platelet Diluent
Description
- Contains Ammonium Oxalate
- For in vitro use only
Specifications
Specifications
| Quantity | 1 x 100 mL |
| For Use With (Application) | For platelet counts, in-vitro diagnostic use only |
| Staining Time | 15 to 20 min. |
| Packaging | Bottle |
| Product Type | Diluting Fluid |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.