Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Potassium Hydroxide Solution (3%)

Catalog No. ES9775
Change view
Click to view available options
Quantity:
1 x 3800 mL (1 gal.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES9775 1 x 3800 mL (1 gal.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES9775 Supplier ENG Scientific Supplier No. 9775
Add to Cart
Add to Cart

Potassium Hydroxide Solution (3%) is a phosphate buffer.

  • Phosphate Buffer

Specifications

Type Potassium Hydroxide
Packaging Type Bottle
Quantity 1 x 3800 mL (1 gal.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.