Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific PPLO Stain

Catalog No. ES6601
Change view
Click to view available options
Quantity:
1 x 100 mL
2 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6601 1 x 100 mL
ES6600 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. ES6601 Supplier ENG Scientific Supplier No. 6601
Add to Cart
Add to Cart
This item is not returnable. View return policy

For staining Mycoplasma colonies. PPLO Stain (for Mycoplasma, Dienes and Weinberger) contains Methylene Blue, Azure II, Maltose, Sodium Carbonate.

  • Mycoplasma, small bacteria without cell walls, are destroyed by the usual methods of smear preparation and heat fixation.
  • On solid culture medium, they form small colonies and are best stained through direct staining of these young colonies.
  • Contents: Methylene Blue, Azure II, Maltose, Sodium Carbonate
  • Solution made from certified dyes

Specifications

Quantity 1 x 100 mL
Certifications/Compliance Made from certified dyes
Format Liquid
For Use With (Application) Staining Mycoplasma colonies
Color Blue
Composition Methylene Blue, Azure II, Maltose, Sodium Carbonate
Volume (Metric) 100 mL
Product Type PPLO Stain
Packaging Bottle

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.