Learn More
ENG Scientific Prussian Blue Stain Kit
Catalog No. ES3160
Change view
Click to view available options
Quantity:
2 x 250 mL
Catalog No. Quantity
ES3160 2 x 250 mL
1 options
This item is not returnable. View return policy
Catalog No. ES3160 Supplier ENG Scientific Supplier No. 3160
Add to Cart
Add to Cart
This item is not returnable. View return policy
For hemosiderin in urine. Course granules of hemosiderin will stain blue. For Prussian Blue Kit.
  • Use as fixative prior to staining specimen material
  • Prussian Blue Stain Kit
  • For in vitro diagnostic use only
  • Solutions I and II
Specifications
Quantity 2 x 250 mL
For Use With (Application) For Hemosiderin in Urine, in-vitro diagnostic use only
Packaging Complete Kit (Bottles)
Product Type Stain Kit
For in vitro diagnostic use only.