Learn More
ENG Scientific Quick Stain Kit, Solution II - Eosin
Catalog No. ES4220
Change view
Click to view available options
Quantity:
1 x 3800 mL
Catalog No. Quantity
ES4220 1 x 3800 mL
1 options
Catalog No. ES4220 Supplier ENG Scientific Supplier No. 4220
Add to Cart
Add to Cart
Quick Stain Kit, Solution II - Eosin for excellent staining results in one minute without precipitate. Alcoholic quick stain technique. No fixation necessary. For Quick Stain Kit (Wright or Wright Giemsa).
  • Yeilds result in one minute
  • No Precipitate
  • Quick Stain Kit
  • Includes Eosin
  • Solution II
  • Use for Alcoholic quick stain technique
  • For in-vitro diagnostic use only
Specifications
Quantity 1 x 3800 mL
For Use With (Application) For Alcoholic quick stain technique, yields result in one min.
Product Type Stain
For in vitro diagnostic use only.