Learn More
ENG Scientific Schiff Reagent
Description
- A stain used in histopathology for the staining of glycogen polysaccharides or in combination with the Periodic Acid-Schiff reaction.
- Made from certified dyes.
- For in vitro diagnostic use only.
Specifications
Specifications
| Content And Storage | Basic Fuchsin, Sodium Sulfite, HCl |
| For Use With (Equipment) | Staining Equipment |
| Product Type | Staining Reagent |
| Quantity | 2 x 250 mL |
| Solution Type | Stain |
| For Use With (Application) | Histopathology for the staining of glycogen polysaccharides or in combination with the Periodic Acid-Schiff reaction |
| Form | Liquid |
| Format | Bottle |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.