Learn More
ENG Scientific Shorr-Single Differential Stain
Description
- Provides a sharp differentiation between cornified and non-cornified elements.
- Made from certified dyes.
- For in vitro diagnostic use only.
- Includes Ethanol, Biebrich Scarlet, Orange G, Fast Green FCF, Phosphotungstic Acid, Phosphomolybdic Acid, Glacial Acetic Acid
Specifications
Specifications
| For Use With (Application) | Provides a sharp differentiation between cornified and non-cornified elements |
| Includes | Ethanol, Biebrich Scarlet, Orange G, Fast Green FCF, Phosphotungstic Acid, Phosphomolybdic Acid, Glacial Acetic Acid |
| Packaging | Bottles |
| Quantity | 2 x 950 mL |
| Product Type | Shorr-single Differential Stain |
For in vitro diagnostic use only.