Learn More
ENG Scientific Solution I - MF Stock Solution
Catalog No. ES6781
Change view
Click to view available options
Quantity:
1 x 250 mL
Catalog No. Quantity
ES6781 1 x 250 mL
1 options
Catalog No. ES6781 Supplier ENG Scientific Supplier No. 6781
Add to Cart
Add to Cart
Solution I - MF Stock Solution is used to achieve stool preservation and staining simultaneously. For MIF Stain and Fixative Kit (Merthiolate-Iodine-Formaldehyde).
  • Solution I - MF Stock Solution
  • Replacement Solution for MIF Stain and Fixative Kit
Specifications
Packaging Bottle
Quantity 1 x 250 mL
Product Type Replacement Solution for MIF Stain and Fixative Kit
For in vitro diagnostic use only.