Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Trichrome Stain Kit, Solution I - PVA Solution (5%)

Catalog No. ES6686
Change view
Click to view available options
Quantity:
1 x 3800 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6686 1 x 3800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ES6686 Supplier ENG Scientific Supplier No. 6686
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For staining of intestinal protozoa. Use with Trichrome Stain Kit (For Intestinal Protozoa) with PVA Solution. Kit contains 100 mL of each Solution. Not to be confused with Gomori's Trichrome Stain used for Histology.

  • For intestinal protozoa
  • With PVA Solution
  • Not to be confused with Gomori's Trichrome Stain Used for Histology

Specifications

For Use With (Application) Staining Intestinal Protozoa
Quantity 1 x 3800 mL
Product Type Replacement Solution for Trichrome Stain Kit

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.