Learn More
ENG Scientific Solution I - Toluidine Blue O Stain
Catalog No. ES7151
Change view
Click to view available options
Quantity:
2 x 250 mL
Catalog No. Quantity
ES7151 2 x 250 mL
1 options
This item is not returnable. View return policy
Catalog No. ES7151 Supplier ENG Scientific Supplier No. 7151
Add to Cart
Add to Cart
This item is not returnable. View return policy
Modified Toluidine Blue O Stain allows the Pneumocystis carinii cysts to be visualized more easily after staining. For Pneumocystis carinii Stain Kit (Modified Toluidine Blue O Stain). Includes: 250 mL each of Solutions I, II, III and IV.
  • Solution I - Toluidine Blue O Stain
  • Replacement solution for Pneumocystis Carinii Stain Kit
  • For staining Pneumocystis Carinii cysts
Specifications
For Use With (Application) Staining Pneumocystis carinii cysts
Packaging Bottles
Quantity 2 x 250 mL
Product Type Replacement solution for Pneumocystis Carinii Stain Kit
For in vitro diagnostic use only.