Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Spore Stain Kit, Solution II - Acetic Acid (5%)

Catalog No. ES6820
Change view
Click to view available options
Quantity:
1 x 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6820 1 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES6820 Supplier ENG Scientific Supplier No. 6820
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solution II - Acetic Acid (5%) is a replacement Solution for the Spore Stain Kit. Contains Basic Fuchsin, Phenol, Ethanol, Acetic Acid, Methylene Blue or Malachite Green.

  • Solution II - Acetic Acid (5%)
  • Replacement Solution for Spore Stain Kit

Specifications

Packaging Bottle
Quantity 1 x 250 mL
Product Type Replacement Solution for Spore Stain Kit

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.