Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Trichrome Stain Kit, Solution II - Iodine Alcohol

Catalog No. ES6691
Change view
Click to view available options
Quantity:
1 x 3800 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6691 1 x 3800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES6691 Supplier ENG Scientific Supplier No. 6691
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For staining of intestinal protozoa; Use with Trichrome Stain Kit (For Intestinal Protozoa) with PVA Solution; Kit contains 100mL of each Solution. Not to be confused with Gomori's Trichrome Stain used for Histology.

  • Solution I - PVA Solution, 5%
  • Iodine Alcohol
  • For staining Intestinal Protozoa

Specifications

For Use With (Application) Ideal for staining of intestinal protozoa
Packaging Bottle
Quantity 1 x 3800 mL
Product Type Iodine Alcohol

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.