Learn More
ENG Scientific Solution IIA - Crystal Violet
Catalog No. ES7162
Change view
Click to view available options
Quantity:
1 x 250 mL
Catalog No. Quantity
ES7162 1 x 250 mL
1 options
Catalog No. ES7162 Supplier ENG Scientific Supplier No. 7162
Add to Cart
Add to Cart
Solution IIA - Crystal Violet for pneumocystis carinii in smears of sputum, transtracheal aspirates, and bronchial washings.
  • Solution IIA - Crystal Violet
  • Replacement solution for Gram-Weigart Stain Kit
  • For staining Pneumocystis Carinii
  • Gram-Weigart Stain Kit
  • Kit includes 100 mL each of Solutions I - 1% Aqueous Eosin Y, Solution IIA - Crystal Violet, Solution IIB - Aniline Oil, Aqueous 2.3%, Solution III - Grams Iodine, Solution IV - Aniline Oil/Xylene and Solution V - Xylene
Specifications
For Use With (Application) Staining Pneumocystis carinii
Packaging Bottle
Quantity 1 x 250 mL
Product Type Replacement solution for Gram-Weigart Stain Kit
For in vitro diagnostic use only.