Learn More
ENG Scientific Spore Stain Kit, Solution IIIA - Brilliant Green
Catalog No. ES6840
Change view
Click to view available options
Quantity:
1 x 250 mL
Catalog No. Quantity
ES6840 1 x 250 mL
1 options
Catalog No. ES6840 Supplier ENG Scientific Supplier No. 6840
Add to Cart
Add to Cart
Solution III - Brilliant Green is a replacement Solution for Spore Stain Kit. Contains Basic Fuchsin, Phenol, Ethanol, Acetic Acid, Methylene Blue or Malachite Green.
  • Solution III - Brilliant Green
  • Replacement Solution for Spore Stain Kit
Specifications
Packaging Bottle
Quantity 1 x 250 mL
Product Type Replacement Solution for Spore Stain Kit
For in vitro diagnostic use only.