Learn More
ENG Scientific Solution III - Concentrated Light Green
Description
Modified Kinyoun Method, which aids in differentiating Crytosporidium oocysts from yeast cells; Specimens may be prepared from fresh material or preserved in 10% buffered formalin
- Modified Kinyoun Method, which aids in differentiating Crytosporidium oocysts from yeast cells
- Specimens may be prepared from fresh material or preserved in 10% buffered formalin
Specifications
Specifications
| Format | Liquid |
| For Use With (Application) | Staining Cryptosporidium oocysts |
| Quantity | 2 x 250 mL |
| Product Type | Solution III - Concentrated Light Green |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.