Learn More
ENG Scientific Solution III - Grams Iodine
Catalog No. ES7164
Change view
Click to view available options
Quantity:
1 x 250 mL
Catalog No. Quantity
ES7164 1 x 250 mL
1 options
Catalog No. ES7164 Supplier ENG Scientific Supplier No. 7164
Add to Cart
Add to Cart
Solution III - Grams Iodine for pneumocystis carinii in smears of sputum, transtracheal aspirates, and bronchial washings.
  • Solution III - Grams Iodine
  • Replacement solution for Gram-Weigart Stain Kit
  • For staining Pneumocystis Carinii
  • Kit includes 100 mL each of Solutions I - 1% Aqueous Eosin Y, Solution IIA - Crystal Violet, Solution IIB - Aniline Oil, Aqueous 2.3%, Solution III - Grams Iodine, Solution IV - Aniline Oil/Xylene and Solution V - Xylene
Specifications
For Use With (Application) Staining Pneumocystis carinii
Packaging Bottle
Quantity 1 x 250 mL
Product Type Replacement solution for Gram-Weigart Stain Kit
For in vitro diagnostic use only.