Learn More
ENG Scientific Solution III - Methylene Blue
Catalog No. ES6923
Change view
Click to view available options
Quantity:
1 x 250 mL
Catalog No. Quantity
ES6923 1 x 250 mL
1 options
This item is not returnable. View return policy
Catalog No. ES6923 Supplier ENG Scientific Supplier No. 6923
Add to Cart
Add to Cart
This item is not returnable. View return policy
Solution III - Methylene Blue is used in staining eosinophils from secretions of the nasal passage, sinuses, bronchi and urinary sediment.
  • For Eosinophil Stain Kit
  • Contains Solution I - Eosin Y, Solution II - Buffer Solution and Solution III - Methylene Blue Solution
Specifications
Packaging Bottle
Quantity 1 x 250 mL
Product Type Methylene Blue
For in vitro diagnostic use only.