Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Solution IIIA - Brilliant Green

Catalog No. ES6522
Change view
Click to view available options
Quantity:
1 x 3800 mL (1 gal.)
2 x 250 mL
2 x 950 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6522 1 x 3800 mL (1 gal.)
ES6520 2 x 250 mL
ES6521 2 x 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ES6522 Supplier ENG Scientific Supplier No. 6522
Add to Cart
Add to Cart

Solution IIIA - Brilliant Green is ideal for staining acid-fast bacteria.

  • Use with Acid-fast stain kit.
  • Modification of the Kinyoun Method.

Specifications

For Use With (Application) Acid-fast stain kit
Quantity 1 x 3800 mL (1 gal.)
Product Type Solution IIIA - Brilliant Green

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.