Learn More
ENG Scientific Solution VII - Xylene
Description
- Solution VII, Xylene
- Staining of Enterocytozoon bienuesi spores
Specifications
Specifications
| Format | Liquid |
| For Use With (Application) | Staining Enterocytozoon bienuesi spores |
| Packaging | Bottles |
| Quantity | 2 x 250 mL |
| Product Type | Solution VII - Xylene |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.