Learn More
ENG Scientific Trichrome Stain Kit, Solution VIII - Xylene
Catalog No. ES6721
Change view
Click to view available options
Quantity:
1 x 3800 mL
Catalog No. Quantity
ES6721 1 x 3800 mL
1 options
This item is not returnable. View return policy
Catalog No. ES6721 Supplier ENG Scientific Supplier No. 6721
Add to Cart
Add to Cart
This item is not returnable. View return policy
For staining of intestinal protozoa. Use with Trichrome Stain Kit (For Intestinal Protozoa) with PVA Solution. Kit contains 100 mL of each Solution; Not to be confused with Gomori's Trichrome Stain used for Histology.
  • Solution VIII - Xylene
  • For staining Intestinal Protozoa
  • Replacement Solution for Trichrome Stain Kit
Specifications
For Use With (Application) Staining Intestinal Protozoa
Packaging Bottle
Quantity 1 x 3800 mL
Product Type Replacement Solution for Trichrome Stain Kit
For in vitro diagnostic use only.