Learn More
ENG Scientific Spore Stain Kit (Loeffler's Methylene Blue)
Catalog No. ES6800
Change view
Click to view available options
Quantity:
3 x 250 mL
Catalog No. Quantity
ES6800 3 x 250 mL
1 options
Catalog No. ES6800 Supplier ENG Scientific Supplier No. 6800
Add to Cart
Add to Cart
Spore Stain Kit (Loeffler's Methylene Blue) is a replacement Solution for the Spore Stain Kit. Contains Basic Fuchsin, Phenol, Ethanol, Acetic Acid, Methylene Blue or Malachite Green.
  • Spore Stain Kit, Loeffler's Methylene Blue
  • Includes Solution I - Basic Carbol Fuchsin; Solution II - Acetic Acid (5%); Solution III - Loeffler's Methylene Blue
Specifications
Includes Solution I - Basic Carbol Fuchsin, Solution II - Acetic Acid (5%), Solution III - Loeffler's Methylene Blue
Quantity 3 x 250 mL
Product Type Stain Kit
For in vitro diagnostic use only.