Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Spore Stain Kit (Brilliant Green)

Catalog No. ES6805
Change view
Click to view available options
Quantity:
3 x 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES6805 3 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ES6805 Supplier ENG Scientific Supplier No. 6805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Spore Stain Kit (Brilliant Green) is a replacement Solution for the Spore Stain Kit. Contains Basic Fuchsin, Phenol, Ethanol, Acetic Acid, Methylene Blue or Malachite Green.

  • Spore Stain Kit
  • Includes 250 mL Solution I - Basic Carbol Fuchsin, 250 mL Solution II - Acetic Acid (5%), 250 mL Solution IIIA - Brilliant Green
  • For In-Vitro diagnostic use only
  • Contents: Basic Fuchsin, Phenol, Ethanol, Acetic Acid, Methylene Blue or Malachite Green
  • 1 Kit

Specifications

Quantity 3 x 250 mL
Includes 250 mL Solution I - Basic Carbol Fuchsin, 250 mL Solution II - Acetic Acid (5%), 250 mL Solution IIIA - Brilliant Green
For Use With (Application) In vitro diagnostic use
Stain Type Spore
Volume (Metric) 250 mL
Product Type Spore Stain Kit
Packaging Complete Kit (Bottles)

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.