Learn More
ENG Scientific Stat-Quick Wright and Wright Giemsa Stain Kits
Description
- Only two solutions needed to obtain excellent staining
- No fixation required with this stain
- With matching buffer solution
- Matching buffer solution allows uniform staining of all cellular elements without precipitate or artifactual changes
- Stain placed in staining container should be mixed periodically
- During cold weather, allow solution to return to room temperature and shake well before use
- Used in both stat and routine laboratories
Specifications
Specifications
| Quantity | 2 x 3800 mL (1 gal.) |
| Packaging | Kit |
| Product Type | Wright Giemsa Stain and Matching Buffer |
For in vitro diagnostic use only.