Learn More
ENG Scientific Sudan IV (Scarlet Red) Stain Kit
Catalog No. ES4900
Change view
Click to view available options
Quantity:
2 x 100 mL
2 x 250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES4900 2 x 250 mL
ES4901 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. ES4900 Supplier ENG Scientific Supplier No. 4900
Add to Cart
Add to Cart
This item is not returnable. View return policy
For microscopic examination of fecal samples for neutral fats.
  • Sudan IV (Scarlet Red) Stain Kits for fat in stool specimens
  • Contains 95 percent ethyl alcohol and Sudan IV
  • Specifically for microscopic examination of fecal samples for neutral fats
  • In vitro diagnostics use only
Specifications
Packaging Complete Kit (Bottles)
Quantity 2 x 250 mL
For in vitro diagnostic use only.