Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Sulfosalicylic Acid 3%

Catalog No. ES9752
Change view
Click to view available options
Quantity:
1 x 3.8 L (1 gal.)
1 x 475 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES9752 1 x 3.8 L (1 gal.)
ES9750 1 x 475 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES9752 Supplier ENG Scientific Supplier No. 9752
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfosalicylic Acid 3%

  • Sulfosalicylic Acid 3%

Specifications

Type Sulfosalicylic Acid
Packaging Type Bottle
Quantity 1 x 3.8 L (1 gal.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.