Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Systain™ Methanol

Catalog No. ES1670
Change view
Click to view available options
Quantity:
1 x 3800 mL (1 gal.)
4 x 3800 mL (1 gal.)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES1670 1 x 3800 mL (1 gal.)
ES1671 4 x 3800 mL (1 gal.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. ES1670 Supplier ENG Scientific Supplier No. 1670
Add to Cart
Add to Cart
This item is not returnable. View return policy

Systain™ Methanol is for use with Sysmex™ hematology slide stainer.

  • Use as fixative prior to staining specimen material
  • In the event of cold weather, allow solutions to return to room temperature

Specifications

Quantity 1 x 3800 mL (1 gal.)
Storage Requirements Room Temperature
For Use With (Application) For in-vitro diagnostic use only, use as fixative
For Use With (Equipment) For Sysmex hematology slide stainer
Packaging Bottle
Product Type Reagent

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.