Learn More
ENG Scientific Systain Wright Stain
Description
- Systain Wright Stain
- For Sysmex hematology slide stainer
Specifications
Specifications
| Quantity | 1 x 3800 mL |
| Storage Requirements | Room Temperature |
| For Use With (Application) | For in-vitro diagnostic use only |
| For Use With (Equipment) | For Sysmex hematology slide stainer |
| Product Type | Stain |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.