Learn More
ENG Scientific Traxstain Buffer
Catalog No. ES1621
Change view
Click to view available options
Quantity:
19 L (5 gal.)
3.8 L (1 gal.)
Product Type:
Traxstain Buffer, pH 6.75
Traxstain Buffer, pH 7.0
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
ES1621 19 L (5 gal.) Traxstain Buffer, pH 7.0
ES1625 3.8 L (1 gal.) Traxstain Buffer, pH 6.75
ES1620 3.8 L (1 gal.) Traxstain Buffer, pH 7.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ES1621 Supplier ENG Scientific Supplier No. 1621
Add to Cart
Add to Cart
For use with EM sciences Midas™ slide stainers, geometric data corporation Hemastainer™ or any other instrument employing a dip stain technique.
  • Traxstain Buffer
  • Compatibile with instruments using a dip stain technique
  • Cubetainer
Specifications
Product Type Traxstain Buffer, pH 7.0
For Use With (Equipment) Instruments using a dip stain technique
Quantity 19 L (5 gal.)
For in vitro diagnostic use only.