Learn More
ENG Scientific Trichrome Stain Kit (with Schaudinn Fixative for PVA Fixative)
Catalog No. ES6682
Change view
Click to view available options
Quantity:
10 x 100 mL
Catalog No. Quantity
ES6682 10 x 100 mL
1 options
Catalog No. ES6682 Supplier ENG Scientific Supplier No. 6682
For staining of intestinal protozoa. Use with Trichrome Stain Kit (For Intestinal Protozoa) with Schaudinn Fixative for PVA Fixative. Kit contains 100 mL of each Solution. Not to be confused with Gomori's Trichrome Stain used for Histology.
  • Trichrome Stain Kit with PVA Solution and Schaudinn Fixative for PVA Fixative
  • Includes Solution I - PVA Solution (5%), Solution IA - Schaudinn Solution, Solution IB - Glacial Acetic Acid, Solution II - Iodine Alcohol, Solution III - Ethanol (70%), Solution IV - Trichrome Stain, Solution V - Acid Alcohol (90%), Solution VI - Ethanol (95%), Solution VII - Carbol-Xylene, Solution VIII - Xylene
  • For staining Intestinal Protozoa
Specifications
For Use With (Application) Staining Intestinal Protozoa
Includes Solution I - PVA Solution (5%); Solution IA - Schaudinn Solution; Solution IB - Glacial Acetic Acid; Solution II - Iodine Alcohol; Solution III - Ethanol (70%); Solution IV - Trichrome Stain; Solution V - Acid Alcohol (90%); Solution VI - Ethanol (95%); Solution VII - Carbol-Xylene; Solution VIII - Xylene
Packaging Bottles
Quantity 10 x 100 mL
Product Type Trichrome Stain Kit with PVA Solution and Schaudinn Fixative for PVA Fixative
For in vitro diagnostic use only.