Learn More
ENG Scientific Unna Stain
Catalog No. ES9621
Change view
Click to view available options
Quantity:
100 mL
2 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES9621 100 mL
ES9620 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES9621 Supplier ENG Scientific Supplier No. 9621
Add to Cart
Add to Cart
Polychromatic stain used in histology for the differentiation of mast cells from plasma cells.
  • Includes Methylene Blue, Ethanol, Potassium Carbonate
  • Made from certified dyes
  • For in vitro diagnostic use only
Specifications
For Use With (Application) Used in histology for the differentiation of mast cells from plasma cells
Includes Methylene Blue, Ethanol, Potassium Carbonate
Packaging Bottle
Quantity 100 mL
Product Type Unna Stain
For in vitro diagnostic use only.