Learn More
ENG Scientific Wayson Stain
Description
- Contains methylene blue, basic fuchsin, phenol, and ethanol
- Bipolar staining is characteristic and is best visualized with Wayson Stain
- Yersinia Pestis organisms stain dark blue, leukocytes stain light blue or purple, and the background stains a light blue
- Wayson Stain
- For identification of Yersinia pestis, in-vitro diagnostic use
Specifications
Specifications
| Quantity | 100 mL |
| Includes | Methylene blue, basic fuchsin, phenol, ethanol |
| Format | Bottle |
| For Use With (Application) | Identification of Yersinia pestis, in-vitro diagnostic use |
| Stain Type | Wayson |
| Volume (Metric) | 100 mL |
| Product Type | Wayson Stain |
For in vitro diagnostic use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.